Learn More
Medchemexpress LLC Streptomycin | 57-92-1 | 99.91% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB1906100MG
Encompass_Preferred
Streptomycin is a potent antibiotic effective against *M. tuberculosis* for tuberculosis (TB) research and various bacterial infections. It functions by strongly binding to nucleic acids, which interferes with and blocks protein synthesis while allowing continued RNA and DNA synthesis. This compound is also commonly used as an antibiotic in culture media and acts as a blocker of stretch-activated and mechanosensitive ion channels in neurons and cardiac myocytes.
  • Effective antibiotic against *M. tuberculosis*.
  • Used for tuberculosis (TB) research.
  • Bactericidal agent for various bacterial infections.
  • Interferes with and blocks protein synthesis.
  • Permits continued RNA and DNA synthesis.
  • Commonly used antibiotic in culture media.
  • Blocks stretch-activated and mechanosensitive ion channels in neurons and cardiac myocytes.
Catalog No. 50-004-11831