Learn More
Research Products International Corp Streptomycin Sulfate 250 G
SDP
Supplier:  Research Products International Corp S62000250.0
Encompass_Preferred
Streptomycin Sulfate is an aminoglycoside antibiotic that inhibits bacterial protein synthesis by binding to the 30S ribosomal subunit, disrupting the initiation complex and leading to misreading of mRNA. This interference prevents proper translation and inhibits bacterial growth. Streptomycin is employed in microbiology for selective bacterial isolation and antibiotic sensitivity testing to assess the susceptibility of bacterial strains. Ideal for studying bacterial susceptibility patterns and antibiotic resistance. Its inclusion in culture media aids in selectively isolating bacteria susceptible to this antibiotic.
Catalog No. 50-297-9109