Learn More
Cayman Chemical StreptozotocIn 1g
SDP
Supplier:  Cayman Chemical 131041 G
Encompass_Preferred
A glucosamine-nitrosourea which is commonly used to induce experimental diabetes in animals; specifically targets beta cells, entering via the glucose transporter GLUT2 and causing alkylation of DNA, leading to the destruction of beta cells
Catalog No. 50-295-5875