Learn More
AdipoGen Streptozotocin,Chemical. CAS: 18883-66-4
SDP
Supplier:  AdipoGen AGCN20046M250
Encompass_Preferred
Streptozotocin, AdipoGen Life Sciences. Chemical. CAS: 18883-66-4. Formula: C8H15N3O7. MW: 265.2. Antibiotic. Diabetogenic. Diabetes inducer. Induces diabetes mellitus in animal models through its toxic effects on pancreatic beta-cells. Mutagenic. Potent alkylating agent. Potent DNA methylating agent. Nitric oxide (NO) donor. Vasorelaxant. Cytotoxic to cells that express GLUT2 glucose transporter. O-GlcNAc-selective N-acetyl-beta-D-glucosaminidase (O-GlcNAcase) inhibitor. Genotoxic. Induces DNA damage. Produces DNA strand breaks. Cell death inducer. Antineoplastic. Anti-cancer agent used in chemotherapy. Induces cell cylce arrest at G2.
Catalog No. 50-464-383