Learn More
Grainger Stretch Wrap Cast Standard Duty
Supplier:  Grainger 15A997
Encompass_Preferred
The Grainger 15C019 Stretch Wrap is a 70 gauge hand film made of linear low density polyethylene. It provides load containment and product protection. This stretch film is available in a 20" x 1500' roll and has 30% pre-stretch. The film is transparent and has an unwind force of 2.5 lbs. It can be used for bundling, unitizing, and palletizing applications. The film adheres to itself and various surfaces.
Catalog No. 50-275-1590