Learn More
IBA LifeSciences StrGat Acptr Vctr pENTRY-IBA51
Supplier:  IBA LifeSciences 54091001
Encompass_Preferred
StarGate Entry Vector is designed to insert the PCR product of a certain gene of interest (GOI) to generate a Donor Vector. StarGate cloning via the pENTRY-IBA vector provides a tool for the systematic and rapid screen of the best working expression conditions for a certain GOI, where nothing is known from literature. After initial pENTRY cloning, the GOI in the resulting Donor Vector can be easily transferred by a simple one-tube reaction into a multitude of expression vectors (Acceptor Vectors) providing different features.
Catalog No. NC3566393