Learn More
IBA LifeSciences StrGate Acptr Vctr pASG-IBA143
Supplier:  IBA LifeSciences 54143001
Encompass_Preferred
The pASG-IBA143 vector is designed for expression of recombinant proteins with an N-terminal His-tag and C-terminal Twin-Strep-tag in E. coli. The vector carries an ampicillin resistance cassette for selection of transformed E. coli cells, F1 and ColE1 origin for a high plasmid copy number, and the inducible tetracycline promoter/operator for the regulated expression of proteins. Expression of the recombinant protein is induced by addition of anhydrotetracycline, which is a derivative of tetracycline that does not exhibit antibiotic activity. In addition to the direct cloning of the gene of interest into pASG-IBA vectors with Esp3I, another option via a so-called Entry Vector is possible.
Catalog No. NC3566401