Learn More
IBA LifeSciences StrGate Acptr Vctr pASG-IBA43
Supplier:  IBA LifeSciences 54043001
Encompass_Preferred
The pASG-IBA43 vector is designed for expression of recombinant proteins with an N-terminal His-tag and a C-terminal Strep-tagII in E. coli. The vector carries an ampicillin resistance cassette for selection of transformed E. coli cells, F1 and ColE1 origin for a high plasmid copy number, and an inducible tetracycline promoter/operator for the regulated expression of proteins. Expression of the recombinant protein is induced by addition of anhydrotetracycline, which is a derivative of tetracycline that does not exhibit antibiotic activity. Please note that cloning into pASG-IBA Acceptor Vectors compulsorily requires the restriction enzyme Esp3I since no other MCS for the integration of a gene of interest is available. In addition to the direct cloning of the gene of interest into pASG-IBA vectors with Esp3I, another option via a so-called Entry Vector is possible.
Catalog No. NC3566356