Learn More
IBA LifeSciences StrGate Acptr Vctr pASG-IBAwt1
Supplier:  IBA LifeSciences 54000001
Encompass_Preferred
The pASG-IBAwt1 vector is designed for expression of target proteins without affinity tag in E. coli. The vector carries an ampicillin resistance cassette for selection of transformed E. coli cells, F1 and ColE1 origin for a high plasmid copy number, and the inducible tetracycline promoter/operator for the regulated expression of proteins. The vector can be combined with any E. coli strain since the tet-promoter works independently of the genetic background of E. coli. Expression of the recombinant protein is induced by addition of anhydrotetracycline, which is a derivative of tetracycline that does not exhibit antibiotic activity. Please note that cloning into pASG-IBA Acceptor Vectors compulsorily requires the restriction enzyme Esp3I since no other MCS for the integration of a gene of interest is available. In addition to the direct cloning of the gene of interest into pASG-IBA vectors with Esp3I, another option via a so-called Entry Vector is possible.
Catalog No. NC3566341