Learn More
IBA LifeSciences StrGate Acptr Vctr pCSG-IBA105
Supplier:  IBA LifeSciences 55105001
Encompass_Preferred
pCSG-IBA105 is a large expression vector with universal features for transient expression of target proteins with an N-terminal Twin-Strep-tag as well as for generation of stable mammalian cell lines. The vector carries an ampicillin resistance cassette for selection of transformed E. coli cells and ColE1 origin for a high plasmid copy number. Extrachromosomal replication in mammalian cells could occur either by origin of replication from Epstein-Barr Virus (oriP) or by SV40 ori. For the former the vector provides the EBNA-1 gene and for the latter the cell line has to be latently infected with SV40 or express the SV40 large T antigen (e.g. HEK293T, COS-1, COS-7). Stable cell lines can be selected by the neomycin resistance gene (NeoR). In addition, the human cytomegalovirus (CMV) immediate-early promoter enables a high-level expression in a wide range of mammalian cells.
Catalog No. NC3566458