Learn More
IBA LifeSciences StrGate Acptr Vctr pESG-IBAwt1
Supplier:  IBA LifeSciences 54400001
Encompass_Preferred
The pESG-IBAwt1 vector is designed for high-level, stable, and non-replicative transient expression of target proteins without affinity tag in mammalian cells. The vector carries an ampicillin resistance cassette for selection of transformed E. coli cells as well as the F1 and ColE1 origin for a high plasmid copy number. In addition, it carries the human cytomegalovirus (CMV) immediate-early promoter for high-level expression in a wide range of mammalian cells, the neomycin resistance gene for selection of stable cell lines, and the SV40 ori for episomal replication in cell lines that are latently infected with SV40 or that express the SV40 large T antigen (e.g. HEK293T, COS-1, COS-7). In addition to the direct cloning of the gene of interest into pESG-IBA vectors with Esp3I, another option via a so-called Entry Vector is possible.
Catalog No. NC3566420