Learn More
Mar Cor Purification Minncare HD Disinfectant 4x1 Quart Container
Supplier:  Mar Cor Purification 3029765
Encompass_Preferred
Minncare HD is a peracetic acid solution developed specifically for use on hemodialysis reverse osmosis (RO) membranes and their associated distribution systems.
Catalog No. NC1972769