Learn More
Enterprise Technology Solutions Stripping Floor Pads, Black
SDP
Supplier:  Enterprise Technology Solutions BWK4020BLA
Encompass_Preferred
For long-lasting stripping of finish, dirt and buildup.For use with rotary machines (Speed of 175-600 rpms).Color-coded for easy job identification.
Catalog No. 50-211-1181