Learn More
Cayman Chemical Strogen RECPTR aB regIon 100uL
SDP
Supplier:  Cayman Chemical 32766100 UL
Encompass_Preferred
An acridone alkaloid active against the chloroquine-sensitive 3D7 and chloroquine-resistant Dd2 strains of P. falciparum (IC50s 29.7 and 33.7 Ug/ml respectively) cytotoxic to PC-3M and LNCaP cancer cells (IC50s 47.9 and 37.8 Ug/ml respectively)
Catalog No. 50-294-6876