Learn More
Strem, An Ascensus Company CAS# 543-94-2. 500g. Strontium acetate, reagent. MFCD00036392
SDP
Supplier:  Strem, An Ascensus Company 933801500G
Encompass
CAS# 543-94-2. 500g. Strontium acetate, reagent. MFCD00036392. Molecular Weight: 205.71. Molecular Formula: Sr(OOCCH3)2. Color/form: white pwdr. Strem# 93-3801. http://www.strem.com/catalog/v/93-3801/
Catalog No. 50-901-15167