Learn More
Medchemexpress LLC Strontium chloride, anhydrous grade | 10476-85-4 | 99.99% | 158.53 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY22009A
Encompass_Preferred
Strontium chloride, anhydrous grade, 99.99% metals basis is a biochemical reagent. This product is suitable for biochemical assays and general laboratory use.
  • High purity, 99.99% metals basis
  • Soluble in water at 100 mg/mL
  • Anhydrous grade
  • Suitable for research use only
Catalog No. 50-004-02263