Learn More
Medchemexpress LLC Strontium chloride, anhydrous grade, 99.99% metals basis | 10476-85-4 | 99.99% | 158.53 | 1 G
SDP
Supplier:  Medchemexpress LLC HY22009A1G
Encompass_Preferred
Strontium chloride, anhydrous grade, 99.99% metals basis is a biochemical reagent. It can be dissolved in H2O at 100 mg/mL (630.80 mM) with the need for ultrasonic assistance. The product should be stored under recommended conditions as specified in the Certificate of Analysis.
  • Biochemical reagent
  • Soluble in H2O at 100 mg/mL (630.80 mM) with ultrasonic assistance
  • Store under recommended conditions as specified in the Certificate of Analysis
Catalog No. 50-004-01148