Learn More
Medchemexpress LLC Strontium-specific chelator | 97759-16-5 | 99.0% | 321.15 | 1 ML

Supplier: Medchemexpress LLC HY1395010MM/1ML
S186 is a strontium-specific chelator designed for research use. It is not intended for patient use.
- A new type of strontium-specific chelator
- For research use only
- Purity of 99.01%
- Appearance as a white to off-white solid
- Molecular weight of 321.15
- Soluble in water at 25 mg/mL (77.85 mM)
- Insoluble or slightly soluble in DMSO
- Solid storage at 4°C, sealed, away from moisture
- In solvent storage at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.