Learn More
Strem, An Ascensus Company CAS# 12036-39-4. 250g. Strontium zirconate, 95%. MFCD00049555
SDP
Supplier:  Strem, An Ascensus Company 933826250G
Encompass
CAS# 12036-39-4. 250g. Strontium zirconate, 95%. MFCD00049555. Molecular Weight: 226.84. Molecular Formula: SrZrO3. Color/form: white pwdr. Strem# 93-3826. http://www.strem.com/catalog/v/93-3826/
Catalog No. 50-901-15192