Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Strophanthidin 10mg | 66-28-4 | 10 MG
SDP

Supplier:  Medchemexpress LLC HY11425210MG

Encompass_Preferred

Strophanthidin is a cardiac glycoside (CAS 66-28-4) supplied for research use. It is used in pharmacological and biochemical studies of Na+/K+-ATPase inhibition and intracellular calcium modulation.

  • High purity (98.53%).
  • Molecular formula C23H32O6 and molecular weight 404.50 g/mol.
  • Available in small research pack sizes suitable for assay development.
  • Soluble in ethanol.
  • Used to study cardiac glycoside activity and Na+/K+-ATPase inhibition.

Catalog No. 50-002-29532


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.