Learn More
Cole-Parmer Stuart 8500 Microcentrifuge, 8500 rpm; 100 to 240 VAC, 50/60 Hz
Supplier:  Cole-Parmer 1741470
Encompass_Preferred
The Stuart 8500 microcentrifuge provides high speed centrifugation with enhanced safety features. The high speed 8500 rpm and 3884 RCF rating allows users to spin samples rapidly or be used in applications where greater g-force is required. Prevent accidental opening with the innovative dual safety locking lid. The unit will only spin when the lid is closed and will automatically shut off after 60 minutes of continuous use. The rubber-coated base allows for enhanced stability and reduced vibration.
Catalog No. 50-196-0430