Learn More
MSC Genesis Ceiling Tile, Vinyl, Revealed Edge, 2 ft x 2 ft, Black
Supplier:  MSC 55663785
Encompass
These ceiling tiles feature a sandstone texture with excellent light reflectance and durability. Designed for a wide range of commercial and institutional settings, they are made from vinyl and come in a black, revealed edge, lay-in design. They are sold in cartons, with each 2x2 tile carton containing 12 tiles.
  • Easy installation, just lays into any grid
  • Waterproof and washable
  • Mold and mildew resistant, ideal for damp or humid spaces
  • Sag resistant
  • Class A fire rating
Catalog No. 50-283-6668