Learn More
Fine Science Tools Student Fine Forceps, Student Stainless Steel, Broad Shanks, 12 cm
SDP
Supplier:  Fine Science Tools 9111310
Encompass
Tip Shape: Broad Shanks/ Tips: Standard/ Tip Dimensions: 0.2 x 0.2mm/ Alloy / Material: Student Stainless Steel/ Length: 11cm/ Feature: Student Quality
Catalog No. NC1045956