Learn More
Fine Science Tools Student Halsted-Mosquito Hemostats, Stainless Steel, 12.5 cm
SDP
Supplier:  Fine Science Tools 9130812
Encompass
Tip Shape: Straight/ Tips: Serrated/ Tip Width: 1.4mm/ Clamping Length: 20mm/ Alloy / Material: Student Stainless Steel/ Length: 12.5cm/ Feature: Student Quality
Catalog No. NC9815172