Learn More
Waters Corp Waters Column, Styragel HMW 2, 500 Å, 20 μm, 7.8 mm x 300 mm
Supplier: Waters Corp WAT054494
Styragel HMW columns are packed with a 20 μm particle and utilize a special 10 μm frit to minimize polymer shearing, making them ideal for use in high-molecular weight applications.
- Separation mode: SEC/GPC
- Support particles: Polymer
- pH range: 2 to 12
- Molecular weight range: 100 to 10000
- Particle shape: Spherical
- Particle size: 20 μm
- Pore size: 500 Å
- Format: Column
- Packing solvent: DMF
- System: HPLC
- Technique: GPC
- Internal diameter: 7.8 mm
- Length: 300 mm
- Range: Styragel
- Product type: Column
- Units per package: 1
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.