Learn More
Waters Corp Waters GPC Column, Styragel HMW 6E, Mixed Bed, 4.6 mm x 300 mm
Supplier: Waters Corp WAT046825
This GPC column is designed for the analysis of ultra-high molecular weight polymers that are prone to shearing. It is a long-life column suitable for various Gel Permeation Chromatography applications.
- Styragel HMW 6E series column
- Particle size: 20 μm
- Dimensions: 4.6 mm I.D. x 300 mm
- Effective molecular weight range: 5,000 to 1 x 107
- Shipped in DMF
- Operating temperature: Ambient to 180 °C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.