Learn More
Waters Corp Styragel HMW 7 Column, 10,000,000 Å, 4.6 mm x 300 mm
Supplier: Waters Corp WAT046805
These narrow bore columns are designed to reduce solvent consumption and disposal costs by up to two-thirds. They are packed with a 20 μm particle and use a special 10 μm frit to minimize polymer shearing, making them suitable for high-molecular weight applications.
- Separation mode: SEC/GPC
- Particle substrate: Polymer
- pH range: 2 to 12
- Molecular weight range: 500,000 to 100,000,000
- Particle shape: Spherical
- Particle size: 15 to 20 μm
- Endfitting type: Waters
- Pore size: 10,000,000 Å
- Format: Column
- Packing solvent: THF
- System: HPLC
- Technique: GPC
- Inner diameter: 4.6 mm
- Length: 300 mm
- Units per package: 1
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.