Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Styragel GPC Column, 15 to 20 μm, 7.8 mm × 300 mm

Supplier:  Waters Corp WAT044200

Encompass_Preferred

Styragel HMW columns are designed for high-molecular weight applications. They feature a 20 μm particle packing and a special 10 μm frit, which work together to minimize polymer shearing during use.

  • Chemistry: GPC
  • Separation mode: SEC/GPC
  • Particle substrate: Polymer
  • PH range: 2 to 12
  • Molecular weight range: 500000 to 100000000
  • Particle shape: Spherical
  • Particle size: 15 to 20 μm
  • Format: Column
  • Packing solvent: Toluene
  • System: HPLC
  • Inner diameter: 7.8 mm
  • Length: 300 mm
  • Units per package: 1

Catalog No. 50-466-771


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.