Learn More
Waters Corp Waters Styragel Column, GPC, 5 μm Particle, 50 Å Pore
Supplier: Waters Corp WAT045840
These narrow bore columns are designed to cut solvent consumption and disposal costs. Styragel HR columns are high-resolution GPC columns, ideal for characterizing samples in the low- to mid-molecular range where high resolution is critical.
- Reduced solvent consumption and disposal costs.
- High-resolution GPC column.
- Ideal for characterizing samples in the low- to mid-molecular range.
- Packed with 5 μm particles.
- Particle substrate: polymer.
- pH range: 2 to 12.
- Molecular weight range: 0 to 1000.
- Spherical particle shape.
- Pore size: 50 Å.
- Packing solvent: DMF.
- System: HPLC.
- Inner diameter: 4.6 mm.
- Length: 300 mm.
- Units per package: 1.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.