Learn More
Waters Corp Waters Corporation Styragel GPC HR2 Column, 5 μm, 500 Å, 4.6 mm x 300 mm
Supplier:  Waters Corp WAT045865
Encompass_Preferred
The Waters Styragel HR2 GPC Column is a high-resolution GPC column with 5 μm particles, ideal for low- to mid-molecular range sample characterization. Its narrow bore design reduces solvent consumption and disposal costs. It operates across a broad 2–12 pH range and is manufactured under stringent ISO, FDA, and cGMP criteria. This column has an effective molecular weight range of 500–20,000 and uses THF as its shipping solvent.
  • High–resolution GPC column packed with 5 μm particles.
  • Ideal for characterizing samples in the low– to mid–molecular range.
  • Narrow bore design reduces solvent consumption and disposal costs by up to two–thirds.
  • Operates across a broad 2–12 pH range.
  • Manufactured following ISO, FDA, and cGMP criteria.
  • Features an effective molecular weight range of 500–20,000.
  • Shipped with THF solvent.
Catalog No. 50-785-592