Learn More
Waters Corp Waters Column, 5 μm, 500 Å, 7.8 mm × 300 mm
Supplier:  Waters Corp WAT044237
Encompass_Preferred
These high-resolution GPC columns are packed with 5 μm particles, ideal for characterizing samples in the low to mid molecular range where high resolution is critical. They are designed for size-exclusion techniques to separate small and large molecules from interfering matrices in various applications.
  • Suitable for applications including foods, pharmaceutical preparations, and natural products.
  • Manufactured under strict conditions to ensure high quality and reliability of results.
  • Adheres to ISO, FDA, and cGMP guidelines.
Catalog No. 50-785-559