Learn More
Waters Corp Waters Corporation GPC Column, 5 μm, 500 Å, 7.8 mm × 300 mm
Supplier: Waters Corp WAT044238
Styragel HR GPC columns are high-resolution columns for characterizing samples in the low to mid molecular weight range. They are packed with 5 μm particles and made of high-performance styrene-divinylbenzene copolymer particles that are totally porous and extremely cross-linked. These columns provide high plate counts for analyzing low-molecular-weight polymers, oligomers, and additives with high resolution.
- Packed with 5 μm particles.
- Pore size of 500 Å.
- Dimensions: 7.8 mm inner diameter × 300 mm length.
- Molecular weight range: 500 to 20000.
- GPC chemistry for size exclusion chromatography.
- High-performance styrene-divinylbenzene copolymer particles.
- pH range from 2 to 12.
- Spherical particle shape.
- Designed for HPLC systems.
- Units per package: 1.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.