Learn More
Waters Corp WATERS Styragel Column, 5 μm, 1000 Å, 4.6 mm × 300 mm
Supplier: Waters Corp WAT045875
These narrow bore columns can cut your solvent consumption and disposal costs by as much as two-thirds. They are high-resolution GPC columns packed with 5 μm particles and are ideal for characterizing samples in the low to mid-molecular range where high resolution is critical.
- Particle size: 5 μm
- Dimensions: 4.6 mm inner diameter, 300 mm length
- Particle substrate: Polymer
- Application: GPC, SEC/GPC, ideal for characterizing samples in the low to mid-molecular range where high resolution is critical
- Chemistry: GPC
- Separation mode: SEC/GPC
- pH range: 2 to 12 pH
- Molecular weight range: 500 to 30000
- Particle shape: Spherical
- Pore size: 1000 Å
- Format: Column
- Packing solvent: Toluene
- System: HPLC
- Units per package: 1
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.