Learn More
Waters Corp Styragel Column, 5 μm, 4.6 mm ID, 300 mm Length
Supplier: Waters Corp WAT045805
These narrow bore columns are designed to reduce solvent consumption and disposal costs while providing high-resolution GPC for characterizing samples in the low to mid-molecular range.
- Reduces solvent consumption and disposal costs.
- High-resolution GPC.
- Packed with 5 μm spherical particles.
- Ideal for low to mid-molecular range characterization.
- Separation mode: SEC/GPC.
- pH range: 2 to 12.
- Molecular weight range: 50 to 100,000.
- Inner diameter: 4.6 mm.
- Length: 300 mm.
- Packing solvent: THF.
- System: HPLC.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.