Learn More
Waters Corp Waters Column, Styragel HR 4E, 5 μm Particle, 7.8 mm ID, 300 mm Length
Supplier:  Waters Corp WAT044240
Encompass_Preferred
Styragel HR (High-Resolution) Columns are designed for analyzing low-molecular-weight samples like oligomers, epoxies, and polymer additives, where high resolution is critical. These GPC columns use rigid 5 μm particles for high resolution and efficiency in the low-to-mid molecular weight range, supporting diverse size-exclusion applications including separation from complex matrices. Manufactured under ISO, FDA, and cGMP guidelines for reliable performance.
  • Engineered for low-molecular-weight sample analysis
  • Optimized for oligomers, epoxies, and polymer additives
  • Features rigid 5 μm particles for high resolution
  • Provides high efficiency in the low-to-mid molecular weight range
  • Supports diverse size-exclusion applications
  • Separates molecules from complex matrices
  • Produced under ISO, FDA, and cGMP guidelines for reliability
Catalog No. 50-785-562