Learn More
Waters Corp Waters Styragel HR 5E Column, 5 μm, 4.6 mm, 300 mm, Mixed Bed, DMF
Supplier:  Waters Corp WAT045825
Encompass_Preferred
Styragel HR 5E narrow bore columns are high-resolution GPC columns packed with 5 μm particles, designed for characterizing samples in the low- to mid-molecular range where high resolution is critical. These columns can significantly reduce solvent consumption and disposal costs by up to two-thirds.
  • Reduces solvent consumption and disposal costs by up to two-thirds.
  • High-resolution GPC columns for accurate characterization.
  • Packed with 5 μm particles for efficient separation.
  • Ideal for low- to mid-molecular range samples requiring high resolution.
Catalog No. 50-785-584