Learn More
Waters Corp Styragel GPC Column, Mixed Bed, 5 μm, 7.8 × 300 mm
Supplier: Waters Corp WAT044227
These high-resolution GPC columns are packed with 5 μm particles and are ideal for characterizing samples in the low to mid molecular range where high resolution is critical. They provide excellent resolution for samples with low molecular weights, enabling analysis of oligomers, epoxies, and polymer additions. Manufactured under stringent ISO, FDA, and cGMP regulations, these columns deliver repeatable results and support applications separating small and large molecules from complex matrices.
- Separation mode: SEC/GPC.
- Polymer particle substrate with spherical shape.
- pH range: 2 to 12.
- Molecular weight range: 2,000 to 4,000,000.
- Particle size: 5 μm.
- Mixed pore size.
- Inner diameter: 7.8 mm.
- Length: 300 mm.
- Packing solvent: Toluene.
- Compatible with HPLC systems.
- 1 unit per package.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.