Learn More
Waters Corp Styragel GPC Column, 5 μm, 7.8 mm X 300 mm, 1,000,000 Å
Supplier: Waters Corp WAT054474
Styragel HR columns are high-resolution GPC columns, featuring 5 μm spherical polymer particles and a 1,000,000 Å pore size. They are optimized for characterizing samples requiring high resolution within the low to mid molecular weight range (200,000 to 10,000,000), utilizing DMF as the packing solvent and compatible with HPLC systems.
- High-resolution GPC applications
- Features 5 μm spherical polymer particles
- Pore size of 1,000,000 Å
- Optimized for samples in the low to mid molecular weight range (200,000 to 10,000,000)
- Uses DMF as packing solvent
- Compatible with HPLC systems
- Operates within a pH range of 2 to 12
- Column inner diameter: 7.8 mm
- Column length: 300 mm
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.