Learn More
Waters Corp Waters Corporation Styragel HT 4 Column, 10 μm, 4.6 mm x 300 mm
Supplier:  Waters Corp WAT045930
Encompass_Preferred
These narrow bore columns are designed to significantly reduce solvent consumption and disposal costs by up to two-thirds.
  • Packed with 10 μm particles with a narrow particle size distribution.
  • Enhances the packed-bed stability within the column.
  • Delivers exceptional column lifetime, particularly in high-temperature GPC applications.
Catalog No. 50-785-604