Learn More
Waters Corp Waters Corporation Column, 10000 Å, 10 μm, 4.6 mm ID, 300 mm
Supplier:  Waters Corp WAT045935
Encompass_Preferred
These narrow bore columns can cut your solvent consumption and disposal costs by as much as two-thirds. The columns are packed with 10 μm particles with a narrow particle size distribution, enhancing the packed-bed stability. These rugged GPC columns deliver exceptional column lifetime in high-temperature GPC applications.
  • Narrow bore columns that can reduce solvent consumption and disposal costs by up to two-thirds
  • Packed with 10 μm particles with a narrow particle size distribution for enhanced packed-bed stability
  • Rugged GPC columns designed for exceptional column lifetime in high-temperature GPC applications
Catalog No. 50-785-605