Learn More
Waters Corp Styragel Column, 10 μm, 100000 Å, 4.6 mm x 300 mm
Supplier: Waters Corp WAT045955
These narrow bore columns are designed for high-temperature GPC applications. They are packed with 10 μm spherical polymer particles, offering enhanced packed-bed stability and exceptional column lifetime.
- Cut solvent consumption and disposal costs by as much as two-thirds.
- Packed with 10 μm particles with a narrow particle size distribution.
- Enhanced packed-bed stability.
- Exceptional column lifetime in high-temperature GPC applications.
- Separation mode: SEC/GPC.
- Particle substrate: Polymer.
- pH range: 2 to 12 pH.
- Molecular weight range: 50000 to 4000000.
- Particle shape: Spherical.
- Pore size: 100000 Å.
- Format: Column.
- Packing solvent: DMF.
- System: HPLC.
- Technique: GPC.
- Inner diameter: 4.6 mm.
- Length: 300 mm.
- Units per package: 1.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.