Learn More
Waters Corp Styragel HR 5 Column, 5 μm, 4.6 mm ID × 300 mm, 50,000–4 × 106 MW Range
Supplier:  Waters Corp WAT045945
Encompass_Preferred
This column is a Styragel HR 5 model, designed for solvent-efficient applications in molecular weight-based separations. It features precise dimensions and a specific particle size to ensure effective chromatographic performance.
  • Suitable for molecular weight range of 50,000–4 × 106
  • Features a 4.6 mm I.D. × 300 mm dimension
  • Equipped with a 5 μm particle size
  • Shipped in toluene solvent
Catalog No. 50-785-607