Learn More
Waters Corp Waters Styragel HT 5 Column, 10 μm, 7.8 mm ID, 300 mm
Supplier: Waters Corp WAT044213
Styragel HT columns are designed for exceptional column lifetime in high-temperature GPC applications. These rugged GPC columns are packed with 10 μm particles with a narrow particle size distribution, enhancing the packed-bed stability.
- Designed for exceptional column lifetime and packed-bed stability in high-temperature GPC applications.
- Features 10 μm spherical polymer particles and a 100000 Å pore size.
- Operates across a 2 to 12 pH range with a molecular weight range of 50000 to 4000000.
- Has an inner diameter of 7.8 mm and a length of 300 mm.
- Packed in THF solvent for use with HPLC systems and GPC technique.
- Supplied as 1 unit per package.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.