Learn More
Waters Corp Waters Styragel HT Column, 10 μm, 100,000 Å, 50,000 to 4,000,000 MW
Supplier: Waters Corp WAT044214
Styragel HT columns feature 10 μm particles with a narrow particle size distribution, which enhances packed-bed stability. These rugged GPC columns are designed for exceptional column lifetime, especially in high-temperature GPC applications.
- Separation type: SEC/GPC
- Particle substrate: Polymer
- pH range: 2 to 12
- Molecular weight range: 50,000 to 4,000,000
- Particle shape: Spherical
- Particle size: 10 μm
- Pore diameter: 100,000 Å
- Format: Column
- Packing solvent: DMF
- System: HPLC
- Technique: GPC
- Inner diameter: 7.8 mm
- Length: 300 mm
- Units per package: 1
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.