Learn More
Waters Corp Waters Corporation Styragel HT 6 Column, 10 μm, 4.6 x 300 mm, 106 Å
Supplier: Waters Corp WAT045960
These narrow bore columns reduce solvent consumption and disposal costs, featuring 10 μm particles for enhanced packed-bed stability. They deliver exceptional column lifetime in high-temperature GPC applications.
- Separation mode: SEC/GPC.
- Particle substrate: Polymer.
- pH range: 2 to 12.
- Molecular weight range: 200,000 to 10,000,000.
- Particle shape: Spherical.
- Particle size: 10 μm.
- Pore size: 106 Å.
- Packing solvent: Toluene.
- System compatibility: HPLC.
- Inner diameter: 4.6 mm.
- Length: 300 mm.
- Units per package: 1.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.