Learn More
Waters Corp Waters GPC Column, 10 μm Particle, 4.6 mm ID, 300 mm Length
Supplier: Waters Corp WAT045970
These GPC columns are designed for ambient and high-temperature analysis of mid-to-high molecular-weight polymers. They are packed with 10 μm particles and can be used with more aggressive solvents, offering exceptional lifetime in high-temperature applications.
- Cuts solvent consumption and disposal costs by up to two-thirds.
- Packed with 10 μm particles with a narrow particle size distribution for enhanced packed-bed stability.
- Suitable for SEC/GPC separation mode.
- Features a polymer particle substrate.
- Operates within a pH range of 2 to 12.
- Effective molecular weight range from 200,000 to 10,000,000.
- Spherical particle shape.
- Pore size of 1,000,000 Å.
- Packed with DMF solvent.
- Compatible with HPLC systems.
- Inner diameter of 4.6 mm.
- Length of 300 mm.
- One unit per package.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.