Learn More
Waters Corp Styragel HT 6 Column, 1,000,000 Å, 10 μm, 7.8 mm × 300 mm
Supplier: Waters Corp WAT044216
Styragel HT columns are packed with 10 μm particles with a narrow particle size distribution, enhancing the packed-bed stability in the column. These rugged GPC columns deliver exceptional column lifetime in high-temperature GPC applications.
- Separation mode: SEC/GPC
- Particle substrate: Polymer
- pH range: 2 to 12
- Molecular weight range: 200,000 to 10,000,000
- Particle shape: Spherical
- Particle size: 10 μm
- Endfitting type: Waters
- Pore size: 1,000,000 Å
- Format: Column
- Packing solvent: THF
- System: HPLC
- Technique: GPC
- Inner diameter: 7.8 mm
- Length: 300 mm
- Units per package: 1 pk
- Hazardous material: This product contains hazardous material which requires special freight handling. Additional charges may apply.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.