Learn More
Waters Corp Waters Column, 10 μm, 4.6 mm, 300 mm, Mixed Bed
Supplier:  Waters Corp WAT045985
Encompass_Preferred
These narrow bore columns can cut solvent consumption and disposal costs by as much as two-thirds. They are packed with 10 μm particles with a narrow particle size distribution, enhancing the packed-bed stability. These rugged GPC columns deliver exceptional column lifetime in high-temperature GPC applications.
  • Narrow bore columns cut solvent consumption and disposal costs by up to two-thirds.
  • Packed with 10 μm particles with a narrow particle size distribution, enhancing packed-bed stability.
  • Rugged GPC columns deliver exceptional column lifetime in high-temperature GPC applications.
Catalog No. 50-785-615