Learn More
Medchemexpress LLC Styrene-divinylbenzene sulfonated copolymer | 69011-20-7 | 98% | 5 G
SDP
Supplier:  Medchemexpress LLC HY468465G
Encompass_Preferred
Styrene-divinylbenzene sulfonated copolymer is a strongly acidic resin. It is a cationic exchange resin made from a microporous styrene/divinylbenzene (DVB) co-polymer with a sulfonic acid group.
  • Strongly acidic resin
  • Cationic exchange resin
  • Made from a microporous styrene/divinylbenzene (DVB) co-polymer
  • Contains a sulfonic acid group
Catalog No. 50-003-58253