Learn More
Apexbio Technology LLC SU 4312 5812-07-7 10mg
SDP
Supplier:  Apexbio Technology LLC B674610
Encompass_Preferred
SU 4312 (CAS 5812-07-7) is a racemic mixture comprising both (Z)- and (E)-isomers each exhibiting distinct receptor tyrosine kinase inhibitory profiles The (Z)-isomer selectively inhibits platelet-derived growth factor receptor (PDGFR) and vascular endothelial growth factor receptor 2 (VEGFR2/FLK-1) while the (E)-isomer targets PDGFR VEGFR2 epidermal growth factor receptor (EGFR) human epidermal growth factor receptor 2 (HER-2) and insulin-like growth factor 1 receptor (IGF-1R) SU 4312 is widely employed in studies investigating tumor angiogenesis cellular signal transduction pathways and the development of targeted therapies in cancer research
Catalog No. 50-000-91848